Learn More
SouthernBiotech™ Neutralite Avidin

Neutralized, deglycosylated avidin purified from egg white
Supplier: SouthernBiotech™ 720006
Alert:
Reagents contain sodium azide. Sodium azide is very toxic if ingested or inhaled. Avoid contact with skin, eyes, or clothing. Wear eye or face protection when handling. If skin or eye contact occurs, wash with copious amounts of water. If ingested or inhaled, contact a physician immediately. Sodium azide yields toxic hydrazoic acid under acidic conditions. Dilute azide-containing compounds in running water before discarding to avoid accumulation of potentially explosive deposits in lead or copper plumbing.
Description
For Research Use Only. Not for Diagnostic or Therapeutic Use.
Flow cytometry; Enzyme-Linked-Immunosorbent-Assay (ELISA); Immunoblotting; Immunohistochemistry
Order Info
To ensure lot-to-lot consistency, each batch of product is tested by flow cytometry, particle concentration, FLISA or ELISA to conform to characteristics of a standard reference reagent.
Specifications
Colorless to Light Brown | |
7.4 | |
Egg White | |
Liquid | |
d-Biotin |
beta-Galactosidase | |
1 mL | |
2°C to 8°C | |
ELISA, Flow Cytometry, Immunoblotting, Immunohistochemistry |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.