Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SouthernBiotech™ Genetically Distinct Collagens: Human Type IV
SDP

Highly purified fibrous protein for a variety of applications

Supplier:  SouthernBiotech™ 125001S

Catalog No. OB1250-01S


Add to Cart

Description

Description

Human origin, Type IV.

  • Can be used as a collagen standard, antigen for antibody production, and coating material in cell culture studies, and in the formation of collagen gels
  • Prepared via controlled and limited pepsin digestion; greater than 90% pure
  • Supplied as a 0.5mg/mL solution in 500mmol/L acetic acid

Specifications

Specifications

0.5 mg/mL
Controlled and limited pepsin digestion, followed by selective salt precipitation
Human
Store at 2°C to 8°C or in small aliquots at -20°C
Cell Culture Coating Material, Antigen for Antibody Production, Collagen Gel Formation, Collagen Standard
0.5 mg
Collagen, Human Type IV
>90% by SDS-PAGE
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit