Learn More
Bioassay Systems QuantiChrom™ Sorbitol Dehydrogenase Assay Kit. For quantitative determination of sorbitol dehydrogenase enzyme activity.
Supplier: Bioassay Systems DSDH100
For quantitative determination of sorbitol dehydrogenase enzyme activity. Key Features: Fast and sensitive. Linear detection range (20 µL sample): 0.1 to 125 U/L for 12 min reaction. Convenient and high-throughput. Homogeneous "mix-incubate-measure" type assay. Can be readily automated on HTS liquid handling systems for processing thousands of samples per day. Method: OD565nm. Samples: Biological samples e.g. plasma, serum, urine, tissue and culture media. Species: All. Procedure: Assay takes 15 min. Kit size: 100 tests. Detection limit: 0.1 U/L.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.