Learn More
CELLTREAT Scientific Products 500mL Solution Bottle, Sterile
SDP
Supplier:  CELLTREAT Scientific Products 229784
Encompass_Preferred
CELLTREAT Scientific Products 500mL Solution Bottle. Compatible with bottle top filters or for use as solution storage containers. Solution bottle neck size is 45mm. Curved bottle design for easy handling. RNase/DNase free and non-pyrogenic. Every csae is printed with the lot number for quality traceability. Polystyrene bottles are packaged sterile, individually wrapped, 24/case.
Catalog No. 50-201-995