Learn More
C Technology Inc Disposable Plastic Vessels
Supplier: C Technology Inc OC00091P50
Single-use plastic sample vessels specifically designed for variable pathlength measurements. These disposable sample vessels support UV-Vis measurements for wavelengths between 220 nm and 800 nm and pathlengths up to 5 mm. Packaged and sold in convenient plastic trays, these single-use disposables help streamline the flow of samples across the SoloVPE system allowing analysts to get accurate results quickly with minimal effort and little or no clean-up.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.