Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Solids Wastewater QC Sample, Ready to Analyze, SPEX CertiPrep™
Solids QC Samples for Wastewater Analysis, 500 mL, SPEX CertiPrep™
Supplier: SPEX CertiPrep 499
Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Description
Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
- Solids QC Samples
- Wastewater analysis
- 500 mL volume
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Specifications
Solids QC Sample | |
Triple-leached LDPE bottle | |
Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis. |
Ready to analyze | |
500 mL | |
AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation |
Safety and Handling
DOTInformation : Nonhazardous
Recommended Storage : Ambient
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Product intended for laboratory use only
Spot an opportunity for improvement?
Provide Content Correction