Learn More
Labconco™ Solid Epoxy Work Surface
SDPSCI_ED
Labconco™ Solid Epoxy Work Surface for use with Protector™ Demonstration Hoods, Protector™ XVS Ventilation Stations, and Protector™ Work Stations.
Supplier:  Labconco™ 3908401/EMD
Catalog No. S28146
Add to cart
Description
Description
  • Molded solid epoxy work surface
  • Chemical-resistant and fire proof
  • Color: Black
  • Dished to contain spills
  • Contoured to fit dimensions of Protector Demonstration Hoods, Protector XVS Ventilation Systems Protector Work Stations
  • No cupsink cutout
  • Offered in 2 ft., 3 ft., 4 ft., 5 ft., 6 ft. and 8 ft. widths and 67.8cm (26.7 in.) and 90.1cm (35.5 in.) depths
Specifications
Specifications
Black
68 cm
1.0 in.
One year
36 in.
91 cm
26.7 in.
Protector™ Demonstration Hoods, Protector™ XVS Ventilation Stations, and Protector™ Work Stations
2.5 m
72 lb.
33 kg
Work Surface