Learn More
LabStrong Corp SOLENOID VALVE PRETREATMENT
Supplier: LabStrong Corp 857X5LS
The LabStrong part number 857X5LS compares to Fi-Streem part number 857X5. The solenoid valve (pretreat, 24V AC) is a replacement part for Fi-Streem Water Purification Stills.