Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Quidel Solana™ STREP Complete Kit
Features rapid method of nucleic acid amplification that does not require thermal cycler. For qualitative detection of Group A β-hemolytic Streptococcus (Streptococcus pyogenes), and pyogenic Group C/G (streptococcus dysgalactiae) nucleic acids.
Supplier: Quidel M921
Includes: Complete Set is comprosed pf one leased Solana Instrument, one Strep Complete Assay Kit, one Controls Set, 95°C Lysis Heat Block, Printer, Labels, Tray, three Racks and Stylus.
Description
- Helicase-dependent amplification (HDA) with fluorescence detection
- Workflow: Easy and flexible, capable of testing a single specimen or batching up to 12 tests at a time
- Intuitive, touch-screen interface with guided operation and customizable settings
- Bench top instrument
- Possesses four USB ports, bar code technology, an external printer
- LIS & middleware capable
- Display: 7 in. touchscreen
- Results reported on screen, stored in the instrument
- Saved to USB drive, printed, and/or sent to the LIS
- Isothermal amplification ensures molecular sensitivity with moderate complexity
- Eliminates thermal cycling to aid in faster time to result (as little as 30 minutes)
- Simplified workflow prevents the need for specialized molecular operator training,
- Small foot-print facilitates decentralization of the instruments
- Sets access rights by operator to prevent unauthorized use or use by untrained operators
- Barcode technology reduces time and errors associated with manually recording patient and operator ID
- Reduce time manually recording test results and risk of transcription errors
- Can connect to many of existing information systems
- Wireless data management and surveillance ecosystem
Order Info
Order eligibility is based upon executed contractual agreement. Please reach out to your Fisher HealthCare Account Representative for more information.
Certifications
CLIA-Moderate
Specifications
Moderate | |
Detection of Group A and pyogenic Group C/G Strep | |
Test, QC, and Calibration results stored on-board |
Molecular Diagnostic Instrument | |
One leased Solana Instrument, one Strep Complete Assay Kit, one Controls Set, 95°C Lysis Heat Block, Printer, Labels, Tray, three Racks and Stylus | |
48 Tests/Kit |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction