Learn More
Quidel Solana™ Group A Strep Starter Kit
A bench top instrument that combines helicase-dependent amplification (HDA) with fluorescence detection to deliver molecular results you can trust, faster than ever before.
Supplier: Quidel M905
Alert:
Refer to Owner's Manual for list of warnings
Includes: Instrument
Heat Block
Printer
Labels
Tray
Racks
Stylus
Description
- Contains Group A Strep Test Kit 48/Pk. and Group A Strep Control Kit
- Flexible workflow: single specimen testing or batching up to 12 tests at a time
- Display: 7″ touchscreen
- Dimensions: 9.4″ X 9.4″ x 5.9″
- Weight: 8.8 lbs
- Isothermal amplification
- LIS and middleware capable
- Barcode Technology
- Offers supervisors the ability to set access rights by operator to prevent unauthorized use or use by untrained operators
- Test, QC, and Calibration results stored on-board
- USB data export to assist in the transfer of data without the need for direct connection
- Optional printing allows for storage of hardcopy results which may enhance post-analytical processes
Order Info
Order eligibility is based upon executed contractual agreement. Please reach out to your Fisher HealthCare Account Representative for more information.
Certifications
CLIA-Moderate
Specifications
Moderate | |
Molecular Diagnostic Instrument | |
Instrument Heat Block Printer Labels Tray Racks Stylus GAS control kit Group A Strep Test Kit 48/Pk. and Group A Strep Control Kit |