Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ICU Medical Sol-Care™ Safety Blood Collection Needle with Multi-Sample Luer
Safety butterfly needle designed to be used for blood collection.
Supplier: ICU Medical 110201030033
Description
- Simple safety mechanism that should be activated immediately after blood withdrawal is complete
- Reduces the risk of needlestick injury
- Thumb rest to ease handling during safety activation
- Flexible wings to secure device to patient's skin
- Thin walled needle is designed to minimize patient discomfort
- Available in 21, 23 or 25 gauge
- Transparent housing and tubing for clear visualization of blood flow
- DEHP-free, flexible, soft, and kink resistant tubing
Specifications
Butterfly Needle with Luer Adapter | |
Sterile | |
Blue | |
23 G | |
19 mm |
PVC | |
Sliding Shield | |
Blood Collection | |
12 in. (tubing) |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction