Learn More
LaMotte™ Soil Texture Unit
For testing and demonstrating different soil textures and properties.
$26.00 - $96.00
Includes: A Study of Soil Science downloadable handbook.
Description
Test soil from schoolyard or samples brought from home and determine the percentage of sand, silt and clay. The overall texture of a soil affects growth in the root zone, which determines the above-ground growth production, and is determined by the fractions of sand, silt, and clay present.