Learn More
Quidel Sofia™ 2 Flu A+B Starter Kit
Small bench top analyzer that uses advanced fluorescence detection with an ultraviolet LED energy source. Sofia 2 Fluorescent Immunoassay Analyzer collects hundreds of data points on the test strip contained inside of a Sofia FIA Test Cassette.
Specifications
Type | Starter Kit |
---|
Alert:
Catalog Number | Mfr. No. | Includes | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Includes | Price | Quantity & Availability | |||||
23-043-089
|
Quidel
20311 |
Sofia 2 Instrument (Fisher Cat 23-043-086) and three Sofia Flu kit (Fisher Cat 23-043-070). |
Pack of 75 for $2,040.00
|
N/A | |||||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
23-043-088
|
Quidel
20310 |
Sofia 2 Instrument (Fisher Cat 23-043-086) and one Sofia Flu kit (Fisher Cat 23-043-070). |
Pack of 25 for $682.50
|
N/A | |||||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
Description
Quidel™ Sofia™ 2 Fluorescent Immunoassay Analyzer (Fisher Cat 23-043-086)
Small bench top analyzer that uses advanced fluorescence detection with an ultraviolet LED energy source. Sofia 2 Fluorescent Immunoassay Analyzer collects hundreds of data points on the test strip contained inside of a Sofia FIA Test Cassette.
- User interface: Intuitive, fast and easy-to-use interface allows operators to quickly run patient samples or navigate stored data
- Objective, automated results: Eliminates operator to operator variability often associated with interpreting visually-read test results. Results are displayed on-screen, printed to an external printer, and/or sent to Virena effortlessly
- Data Management: Results can be reported and stored in multiple ways to meet your data management needs:
- USB storage provides PC storage and analysis capabilities
- Optional printing on an external printer allows for storage of hardcopy results which may enhance post-analytical processes
- Test, QC, and Calibration results stored on-board
- Virena wireless connectivity
- Fluorescent technology: Optimized performance
- Two development modes: Offers flexibility by accommodating specific workflow demands
- Walk away automation: No need for operator to wait for results
- Security: Offers supervisors the ability to set access rights by operator to prevent unauthorized use or use by untrained operators
- Barcode technology: Reduce time and errors associated with manually recording patient and operator ID
- Virena™ Connectivity: Reduce time manually recording test results
- Li Po rechargeable Battery: Can be used anywhere testing needs to be done
- Dimensions (D x W x H): 22 x 12 x 12cm
- Weight: 2 lb. (1kg)
- Power supply input:100-240 VAC, 50-60 Hz, self-switching (U.S./ International), 2.93
- Analyzer input: 7.5V DC 2.5A
- Replacement battery: Li Po rechargeable
- Display: 4-inch color LCD touchscreen
- LAN interface: RJ45 connector
- Operating temperature:15°C to 30°C/59°F to 86°F
- Operating humidity: 20% to 85% RH non condensing
- Operating altitude: 2000 m maximum
- Shipping and storage temperature: −20°C to 55°C
- Shipping and storage humidity: 20% to 85% RH non condensing
- Patient test result storage: 1000 tests maximum
- QC results storage: 200 results maximum
- Calibration results storage: 200 results maximum
Quidel™ Sofia™ Influenza A+B FIA Kit (Fisher Cat 23-043-070)
For rapid differential diagnosis of acute influenza A and influenza B viral infections- CLIA Complexity: Waived for Nasal and Nasopharyngeal swab specimens
- CPT codes: 87804/87804-59
- Detects and differentiates influenza A and B viral infections
- For use with Sofia Analyzer
- Results in 15 minutes
- Enhanced assay sensitivity
- In vitro diagnostic use only
- Employs immunofluorescence
- Detect influenza nucleoprotein antigens in nasal swab and nasopharyngeal swab and NPA wash samples from symptomatic patients
Specifications
Starter Kit |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.