Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sodium Pentacyanoammine Ferroate, MP Biomedicals
Click to view available options
Quantity:
10 g
Specifications
Quantity | 10 g |
Formula Weight | 266.99 |
Chemical Name or Material | Sodium Pentacyanoammine Ferroate |
Safety and Handling
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction