Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fisher Science Education™ Sodium Nitrate
![SCI_ED](/content/dam/fishersci/glyphs/scied.png)
Click to view available options
Quantity:
100 g
500 g
2 kg
Packaging:
Lab Grade
Reagent Grade
Description
This product is for educational use - no C of A is available.
Specifications
Color | White |
Quantity | 500 g |
Assay Percent Range | >95% |
Linear Formula | NaNO3 |
Formula Weight | 84.99 |
Grade | Laboratory |
Packaging | Lab Grade |
Chemical Name or Material | Sodium Nitrate |
Safety and Handling
![](http://beta-static.fishersci.com/images/Yellow.gif)
ShelfLife | 5 years |
DOTInformation | DOT Class 5.1, III, : Oxidizer |
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction