Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sodium Hydroxide (NF/EP/BP/FCC), Fisher Chemical™
Click to view available options
Quantity:
500 g
1 kg
2.5 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Specifications
pH | 14 |
Color | White |
Boiling Point | 1390°C |
Melting Point | 318°C |
UN Number | 1824 |
Density | 2.13g/cm³ |
Quantity | 2.5 kg |
Formula Weight | 40 |
Percent Purity | 95.0 to 100.5% (FCC) |
Grade | BP/EP/FCC/NF |
Show More |
Safety and Handling
Hazard Statement | DANGER! |
Precautionary Statement | Emergency Overview Causes severe burns by all exposure routes. Hygroscopic. Use personal protective equipment. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Move to fresh air. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Immediate medical attention is required. Immediate medical attention is required. Do not induce vomiting. NFPA Health:3 Flammability:0 Instability:1 |
DOTInformation | DOT Class 8, : Corrosive |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction