Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium fluorophosphate, Thermo Scientific Chemicals

Click to view available options
Quantity:
500 g
10163-15-2
FNa2O3P
143.949
MFCD00014248
BFDWBSRJQZPEEB-UHFFFAOYSA-L
sodium monofluorophosphate, disodium monofluorophosphate, sodium fluorophosphate, disodium fluorophosphate, phosphorofluoridic acid, disodium salt, sodium phosphorofluoridate, disodium phosphorofluoridate, extra-strength aim, sodium fluorophosphate na2po3f, unii-c810jcz56q
24266
CHEBI:86431
disodium;fluoro-dioxido-oxo-$l^{5}-phosphane
[O-]P(=O)([O-])F.[Na+].[Na+]
10163-15-2
FNa2O3P
143.949
MFCD00014248
BFDWBSRJQZPEEB-UHFFFAOYSA-L
sodium monofluorophosphate, disodium monofluorophosphate, sodium fluorophosphate, disodium fluorophosphate, phosphorofluoridic acid, disodium salt, sodium phosphorofluoridate, disodium phosphorofluoridate, extra-strength aim, sodium fluorophosphate na2po3f, unii-c810jcz56q
24266
CHEBI:86431
disodium;fluoro-dioxido-oxo-$l^{5}-phosphane
[O-]P(=O)([O-])F.[Na+].[Na+]

 

Sodium fluorophosphate is used as an anti-inflammatory, glucocorticoid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Physical Form Powder
Quantity 500 g
UN Number UN3288
Solubility Information Soluble in water. Insoluble in ethanol and ether.
Formula Weight 143.95
Odor Odorless
Chemical Name or Material Sodium fluorophosphate

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P264b-P270-P301+P312-P330-P501c

H302

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLID, INORGANIC, N.O.S.

EINECSNumber : 233-433-0

RTECSNumber : TE6130000

TSCA : No

Recommended Storage : Ambient temperatures

RUO – Research Use Only