Learn More
Hanna Pharmaceutical Supply Co., Inc. 0.9% SODIUM CHLOR.IV BAG USP 100ML
Supplier: Hanna Pharmaceutical Supply Co., Inc. BRAUNS80045264
Sodium Chloride Injections USP are sterile, nonpyrogenic, isotonic and contain no bacteriostatic or antimicrobial agents These intravenous solutions are indicated for use in adults and pediatric patients as sources of electrolytes and water for hydration The PAB container is PVC-Free and DEHP-Free The plastic container is a copolymer of ethylene and propylene formulated and developed for parenteral drugs The copolymer contains no plasticizers and exhibits virtually no leachability The container has two ports, one is for the intravenous administration set and the other is a medication addition site Each is covered by a tamperproof barrier This product is designed for use as a diluent and delivery system for intermittent intravenous administration of compatible drug additives 64/CS
Please note: This item is nonrefundable, non-returnable and non-cancelable once order is placed.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.