Learn More
Sodium Ascorbate, USP, bioCERTIFIED™, 2.5 kg, Spectrum Chemical
Description
bioCERTIFIED™ products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Spectrum Chemical's bioCERTIFIED™ quality management program goes beyond standard testing and includes APIs, excipients and many more chemicals that carry additional certifications and documentation specifically designed to meet the needs of biopharmaceutical businesses including manufacturers and pharmacies. Each bioCERTIFIED™ chemical undergoes expanded testing for bioburden, endotoxin, elemental impurities and other tests required by the monographs.
Specifications
Quantity | 2.5 kg |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.