Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Desco™ Snap-One™ Dual-Wire Coil Cord with 4 mm Snaps
Dual Conductor Cord designed to maintain the path-to-ground through the other conductor if one is damaged.
Supplier: Desco™ 19696
Description
- 3.5 mm right angle mono plug
- Fits standard dual wire monitors and grounding jacks
- Park-it hole provides a park station for the mono plug a cord when not in use
- Date code provided so that the lifespan of the product may tracked and to ensure quality control
Specifications
PVC | |
Black | |
12 ft. | |
3.66 m |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction