Learn More
Motic SMZ171-TLED Stereo and Camera Bundle
The SMZ-171 is an advanced stereo microscope for biomedical work and industrial quality control.
Supplier: Motic 1100501300051
This item is not returnable.
View return policy
Description
- Greenough Zoom Optical System
- 45° stereomicroscope head
- 6.7:1 Zoom Ratio
- Diopter Adjustment: ±5
Specifications
10X | |
SMZ-171-TLED with BMH4000 Camera Bundle | |
Moticam BMH4000 | |
WF 10X/23 mm |
Auxiliary Objectives (0.3X, 0.5X, 0.63X, 0.75X, 1.5X, 2X) | |
Trinocular | |
LED |
Safety and Handling
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov