Learn More
Smiths Medical Pulsator™ Plus Dry Heparin Arterial Blood Sampling Kits
Pulsator™ Plus 3 mL arterial blood sampling syringes utilize lithium heparin with a concentration of 23.5 I.U. per mL. The specially formulated dry lithium heparin provides the necessary anticoagulation along with minimal interference with electrolyte values, for more accurate and consistent results. The syringes are available with a choice of product features.
$460.00 - $499.00
Specifications
For Use With (Application) | Provides the necessary anticoagulation along with minimal interference for more accurate and consistent results, rapid fill minimizes patient discomfort |
---|---|
Volume (Metric) | 3 mL |
Includes: 3-cc or 5-cc Luer lock syringe and Filter-Pro™ device in polybag or Tyvek packaging.
Catalog Number | Mfr. No. | Product Type | Quantity | Packaging | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Product Type | Quantity | Packaging | Price | Quantity & Availability | |||
22-024-975
|
Smiths Medical
4044P1 |
With Accessory Kit | 100/Cs. | Sterile Tyvek™ |
Case of 100 for $467.50
|
|
22-024-976
|
Smiths Medical
40462 |
Without Needle | 200/Cs. | Polybag |
Case of 200 for $499.00
|
|
22-024-988
|
Smiths Medical
4080P2 |
With patient label, adhesive bandage | 200/Cs. | Sterile Tyvek™ |
Case of 200 for $359.50
|
N/A |
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
22-024-949
|
Smiths Medical
4084P1 |
With Second Needle, Accessory Kit | 100/Cs. | Sterile Tyvek™ |
Case of 100 for $460.00
|
|
Description
- Pulsating effect allows for precise positioning in arterial lumen
- Syringe response to arterial pressure helps ensure arterial sample
- Rapid fill minimizes patient discomfort
- 3mL arterial blood-sampling syringes are easily aspirated during low blood pressure or code situation
- Needles with different gauges and lengths
- Utilize lithium heparin with a concentration of 23.5 IU/mL
- Specially formulated dry lithium heparin provides the necessary anticoagulation along with minimal interference for more accurate and consistent results
Order Info
- With or without support kit (alcohol prep pad, iodophor prep pad, gauze pad, patient label, biohazard labeled ice bag, adhesive bandage)
- Sterile Tyvek™ packaging
- Come with Filter-Pro™ air bubble removal device
Specifications
Provides the necessary anticoagulation along with minimal interference for more accurate and consistent results, rapid fill minimizes patient discomfort | |
3 mL |
Safety and Handling
ShelfLife : 3 Years
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.