Learn More
ICU Medical Pro-Vent™ Plus Arterial Blood Gas Sampling 1cc/23.5IU Kits
Pro-Vent™ Plus 1 mL Arterial Blood Sampling syringes utilize lithium heparin with a concentration of 23.5 I.U. per mL. The specially formulated dry lithium heparin provides the necessary anticoagulation along with minimal interference for more accurate and consistent results.The syringes are available with a broad choice of product features.
Specifications
For Use With (Application) | Provides the necessary anticoagulation along with minimal interference for more accurate and consistent results |
---|---|
Packaging | Sterile Tyvek™ |
Volume (Metric) | 1 mL |
Sterility | Sterile |
Catalog Number | Mfr. No. | Quantity | Includes | Needle Gauge | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Includes | Needle Gauge | Price | Quantity & Availability | |||
22-004-204
|
ICU Medical
4610P2 |
200/Cs. | 1mL luer-slip syringe with Filter-Pro™ device, 25G x 0.62 in. preattached needle, and Point-Lok™ device, 23.5 IU/mL concentration of dry lithium heparin as anticoagulant | 25 G |
Case of 200 for $602.00
|
N/A | |||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
22-004-187
|
ICU Medical
4611P1 |
100/Cs. | 1mL luer-slip syringe with Filter-Pro™ device, 25G x 0.62 in. preattached needle, Point-Lok™ device, support kit (Alcohol prep pad, gauze pad, patient label, biohazard labeled ice bag, adhesive bandage) | 25 G |
Case of 100 for $513.50
|
N/A | |||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
22-004-186
|
ICU Medical
4617P1 |
100/Cs. | 1mL luer-slip syringe with Filter-Pro™ device, 25G x 0.62 in. preattached needle with Needle-Pro™ and support kit (Alcohol prep pad, gauze pad, patient label, biohazard labeled ice bag, adhesive bandage) | 25 G |
Case of 100 for $309.50
|
N/A | |||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
22-004-185
|
ICU Medical
4619P2 |
200/Cs. | 1mL luer-slip syringe with Filter-Pro™ device, and 25G x 0.62 in. preattached needle with Needle-Pro™ needle safety device, 23.5 IU/mL concentration of dry lithium heparin as anticoagulant | 25 G |
Case of 200 for $658.50
|
N/A | |||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
22-004-184
|
ICU Medical
4620P1 |
100/Cs. | 1mL luer-slip syringe with Filter-Pro™ device, 25G x 0.62 in. preattached needle with Needle-Pro™ needle safety device, support kit (Alcohol prep pad, gauze pad, patient label, biohazard labeled ice bag, adhesive bandag | 25 G |
Case of 100 for $383.50
|
N/A | |||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
22-004-201
|
ICU Medical
4640P1 |
100/Cs. | 1mL luer-slip syringe with Filter-Pro™ device, 25G x 0.62 in. preattached needle with Needle-Pro™ needle safety device, 23 G x 1 in. secondary needle with Needle-Pro™ needle safety device attached | 23 to 25 G |
Case of 100 for $446.50
|
N/A | |||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
Description
- Advanced-formula dry lithium heparin
- Offsets binding and dilution factors by artificially pre-adjusting each electrolyte and metabolite to its normal blood level
- Consistent and accurate readings for pH, pCO2, pO2, Ca2+ , Mg2+ , Na+, K+, Cl-, CO-oximetry, glucose, lactate
- Large venting filter media facilitates quick filling and the siliconized barrel allows easy aspiration, even in code situations
- Filter-Pro™ device permits ease of removing air bubbles without accidentally expelling blood or aerosolizing the sample
- Needle-Pro™ needle protection device (sturdy, puncture-resistant material) lock in place, completely covering used needle
- Support kits contain alcohol prep pad, lodophor prep pad, gauze pad, patient label, biohazard-labeled ice bag, adhesive bandage
Order Info
Sterile Tyvek packaging except as noted.
Specifications
Provides the necessary anticoagulation along with minimal interference for more accurate and consistent results | |
Sterile Tyvek™ | |
1 mL | |
Sterile |
Safety and Handling
ShelfLife : 3 Years
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.