Learn More
ICU Medical Anaerobic Pulsator™ Plus Arterial Blood Sampling Kit
Pulsator™ Plus syringes, with a liquid sodium heparin concentration of 23.5 I.U. per mL, allowing blood sampling with minimal risk of sample dilution. The syringes use a specially formulated electrolyte-balanced liquid sodium heparin that offsets both binding and dilution factors by chemically pre-adjusting key electrolytes to normal blood levels. The syringes are available with a broad choice of product features.
Supplier:  ICU Medical 4068NP2
 View more versions of this product
Catalog No. 22-024-986
Item Discontinued Due to product restrictions, please to purchase or view availability for this product.This item has been discontinued by the supplier.
Explore available promotions
Description
Description
Anaerobic Pulsator Plus syringes, with a liquid sodium heparin concentration of 23.5 IU/mL, allow for clot-free blood sampling without risking sample dilution. Specially formulated electrolyte-balanced liquid sodium heparin offsets both binding and dilution factors by chemically pre-adjusting key electrolytes to normal blood levels, providing consistent and accurate analysis for all parameters tested.
  • Consistent and accurate readings of pH, pOC2, pO2, Ca2+, Mg2+ , Na+, K+, Cl-, CO-oximetry, glucose, and lactateFill rapidly, to reduce fill time and minimize patient discomfort
  • Pulsating effect allows for precise positioning in arterial lumen
  • Syringe can be easily aspirated during a low blood pressure or code situation
  • Patented mini-volume insert needle and hub has a residual liquid heparin dead space of 0.03 mL; allows liquid heparin to be thoroughly expelled from syringe and needle, leaves behind an accurate trace coating of heparin
  • Needles available with different gauges, lengths and safety devices (Needle-Pro™, Point-Lok™ or Filter-Pro™)
  • With or without support kit (alcohol prep pad, iodophor prep pad, two gauze pads, patient label, biohazard labeled ice bag, adhesive bandage)
Order Info
  • Needles available with different gauges, lengths and safety devices (Needle-Pro™, Point-Lok™ or Filter-Pro™)
  • With or without support kit (alcohol prep pad, iodophor prep pad, two gauze pads, patient label, biohazard labeled ice bag, adhesive bandage)
Specifications
Specifications
22 G
3mL luer-lock syringe with Filter-Pro™ device, 22G x 1 in. needle with separate Needle-Pro™ needle safety device, mini-volume insert needle, 23.5 IU/mL concentration of liquid sodium heparin
200/Cs.
3 mL
Allow for clot-free blood sampling without risking sample dilution, offsets both binding and dilution factors by chemically pre-adjusting key electrolytes to normal blood levels, providing consistent and accurate analysis for all parameters tested
Arterial Blood Sampling Kit
Polybag
Sterile
Safety and Handling
Safety and Handling
ShelfLife : 3 Years