Learn More
HORIZON DISCOVERY BIOSCIENCES LIMITED SMARTVECTOR IND HU CNOT6 PKG
Supplier:  HORIZON DISCOVERY BIOSCIENCES LIMITED V3SH767513EG57472
Encompass
Set of 3 SMARTvector Inducible Human CNOT6 PGK-TurboRFP shRNA, 100 μL, 10^7 TU/mL
Catalog No. NC2226117