Learn More
Perkin Elmer US LLC SMARTintro Geological Sample Introduction Kit - Gold
Supplier: Perkin Elmer US LLC N8150040
PerkinElmer introduces the NexION 1000/2000 ICP-MS solution-ready sample introduction technology. Engineered for the analysis of geochemical samples containing HF-resistant system utilizes a PFA nebulizer, sapphire injector, and quartz torch to deliver absolute accuracy and precision. This kit includes: PeriPump Tubing Starter Kit (N8150060) PFA Spray Chamber with PFA End Cap (N8150450) Plasma Light Shield (N8152377) PFA Nebulizer (N8152378) Demountable Quartz Torch (N8152379) Internal Standard Addition Tee for PFA ST Nebulizer (N8152423) Torch Injector Nut Removal Tool (N8152443) Sapphire 2 mm Injector (N8152447) Gas Line Addition (N8152449) Torch Injector Nut (N8152451) Internal Standard Probe Qty. 2 (N8152452) Sample Introduction Line for PFA-ST Nebulizer (N8152455) Spray Chamber Drain Line 1/4-28 Fitting (N8152456) PC3 Spray Chamber Light Shield (M8152475) Cassette Only Gold (N8152532)
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.