Learn More
Eppendorf™ SmartExtender Heating Rack PROMO
Add-on for thermoblocks and thermomixers that provides additional/parallel incubation wells.
Supplier:  Eppendorf™ 2231001047
Catalog No. 05-414-307PM
Add to Cart
Description
Description
  • Can easily be used as an add-on to your existing Eppendorf™ SmartBlocks and the Eppendorf™ ThermoMixer C/Fx as well as Eppendorf™ ThermoStat C
  • Up to 12 additional vessels (1.5 mL) can be heated up for higher throughput and greater flexibility
  • Independent heating function for the second temperature enables a parallel usage of an Eppendorf™ SmartBlock and the SmartExtender
  • The Eppendorf™ SmartBlock at the center can still be used for heating, cooling, and mixing
  • Easy, tool-free installation in seconds
  • Active heating only
  • Compatible to all Eppendorf™ SmartBlocks
  • Firmware update required for use
TRUSTED_SUSTAINABILITY
Specifications
Specifications
25°C to 110°C
SmartBlocks, ThermoMixer™ C/Fx, ThermoStat C
12 x 1.5 mL
Product Suggestions
Product Suggestions