Learn More
Cole-Parmer Smart Spatula 17211 Disposable Spatula, blue 210 mm
Supplier:  Cole-Parmer 0626518
Encompass_Preferred
Disposable polypropylene (PP) spatulas are ideal for chemical and biological material transfer, collection, and manipulation. Spatulas features both a scoop end and a spatula end. The contoured spatula end is shatter resistant even when exposed to liquid nitrogen and is flexible enough to fit into many small containers to scrape or collect samples. The hollow core reduces heat and cold transfer. Can be used with dilute acids and bases. Autoclavable.
Catalog No. NC0133733