Learn More
Omron Healthcare, Inc Small D-Ring Cuff 7 to 9 Advanced Accuracy Series
Supplier: Omron Healthcare, Inc CDCS9
Omron charges a $35 drop ship fee on every order. The durable D ring contoured cuff fits arms 7 to 9 inch in circumference. Before you purchase a home blood pressure monitor, it's important to determine the best cuff size for you. To determine your arm size, use a cloth measuring tape and measure the circumference of your left upper arm midway between your elbow and shoulder. Ensuring that the blood pressure cuff is neither too tight nor too loose will help ensure the most accurate readings providing you a more accurate picture of your heart's health.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.