Learn More
IMEB, Inc Pathology Bone Band Saw Optional Small Bone Holder
Supplier: IMEB, Inc SH352
Small Bone Holder for IMEB bone band saw (Pictured femoral head holder and small bone holder). Bone Band Saw sold separately. The IMEB, Inc. Bone Band Saw is an efficient and economical way of cutting bone in laboratory. The bone band saw is only 19" high and has a foot print of 17" x 13" of counter space. This saw has a separate water-cooled blade that eliminates dust and particles.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.