Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Kord™ Slippable Petri Dishes
For standard microbiology and general laboratory use.
Supplier: Kord™ 2914
Description
- Slippable rimmed lids provide semi-stacking feature
- Deep dish design in mono style
- Polystyrene construction
- With ISO mark target
Specifications
Slippable | |
Single-use | |
Round | |
Slippable Petri Dish | |
Standard microbiology and general laboratory use | |
15 mm |
Media saver | |
Polystyrene | |
Sterile | |
1 | |
25/Sleeve | |
95 mm |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction