Learn More
Strategic Applications Inc 600ul K3 EDTA Blood Tube, Slip Tip, 100/pk
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Strategic Applications Inc MULE600
![Encompass](/content/dam/fishersci/glyphs/encompass.png)
600uL K3 EDTA Blood Collection Tube for collection of blood in preclinical studies via capillary action or venipuncture. 100/box. Product is for laboratory animal research only. ** If a COA is required it must be requested when the order is placed or a $50.00 fee will be apply. Contact Thermo Fisher Customer Service at 800-766-7000 to request to COA. **
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.