Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
3M™ Safety-Walk™ Slip Resistant General Purpose Tape, Series 600
Mineral coated, slip resistant material features a highly durable surface. Low profile design helps reduce tripping hazards.
$118.00 - $157.00
Specifications
Adhesive Type | Pressure Sensitive Adhesive |
---|---|
Finish | Mineral-coated |
For Use With (Application) | Light to Heavy Traffic Areas |
Length (Metric) | 18.3 m |
Catalog Number | Mfr. No. | Width (Metric) | Color | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Width (Metric) | Color | Price | Quantity & Availability | ||||
17-623-919
|
3M™ Safety-Walk™
7100032939 |
19 mm | Black |
Case of 4 for $118.00
|
|
||||
17-623-928
|
3M™ Safety-Walk™
7100067526 |
25 mm | Black |
Case of 4 for $157.00
|
|
||||
17-623-861
|
3M™ Safety-Walk™
7000052247 |
25 mm | Yellow |
Case of 4 for $157.00
|
|
||||
17-623-910
|
3M™ Safety-Walk™
7100000852 |
51 mm | Yellow |
Case of 2 for $157.00
|
|
||||
Description
- Anti-slip tape for light to heavy shoe-traffic areas
- High quality abrasive particles to ensure long-lasting friction in any application
- Highly durable surface withstands the rigors of equipment traffic
- High performance, pressure-sensitive adhesive for easy application
- Custom shapes and sizes are also available
- NFSI High Traction Certification
Specifications
Pressure Sensitive Adhesive | |
Mineral-coated | |
Light to Heavy Traffic Areas | |
18.3 m |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title