Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Accuform™ Slip-Gard™ Floor Marking Kit: CAUTION KEEP AREA IN FRONT OF ELECTRICAL PANEL CLEAR FOR 36 INCHES KEEP CLEAR
Pre-cut floor sign kits help keep areas in front of electrical panels free and clear of obstructions.
Supplier: Accuform™ PSR526
Description
- Constructed of thick adhesive vinyl, subsurface printed, with clear laminate to protect the words and image
- Non-slip, pebble-textured surface provides added durability to withstand heavy pedestrian traffic
- Strong, permanent, self-adhesive underside has peel-off back liner
- Adheres firmly to smooth, clean, dry floor surfaces
- Kit includes three pre-cut borders strips: two 2 x 36 in., one 2 x 24 in., and one 17 in. round floor sign (covers a 36 x 24 in. area)
Specifications
Non-glare | |
1 Sign | |
Pressure-sensitive | |
Rectangular | |
CAUTION KEEP AREA IN FRONT OF ELECTRICAL PANEL CLEAR FOR 36 INCHES KEEP CLEAR | |
Black, Yellow |
Laminated Polyester | |
36 x 24 in. | |
Self-adhesive | |
English | |
Electrical Hazard Sign |
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction