Learn More
Newcomer Supply Periodic Acid Schiff (PAS) Glycogen Control Slides, 10/set
Supplier: Newcomer Supply 4540A
Newcomer Supply Periodic Acid Schiff (PAS) Glycogen Control Slides are for the positive histochemical staining of glycogen in tissue sections and can also be utilized as control slides for glycogen digestion steps. Periodic Acid Schiff (PAS) Glycogen Control Slides contain sections of positive staining liver.