Learn More
Epredia™ Slide Baskets and Hangers for Epredia™ ClearVue™ Coverslipper
Use these slide baskets and hangers with the Epredia™ ClearVue™ Coverslipper to make it compatible with any automated slide stainer.
Supplier: Epredia™ A79210066
Description
- Allow equipment compatibility with a variety of different stainers
- Can be loaded in mix-or-match fashion
- Sold in sets of five
Specifications
Coverslipper Basket/Hanger | |
Epredia ClearVue Coverslipper | |
Sakura DRS 2000-compatible basket, black sliders | |
5 Pk. |