Learn More
Fine Science Tools Bone Pliers, Stainless Steel, Straight, 15.5 cm
Supplier: Fine Science Tools 1602514
Tip Shape: Straight/ Alloy / Material: Stainless Steel/ Length: 15.5cm/ Joint Action: Single