Learn More
Med Vet International SKIN ADHESIVEDERMABOND12BX
Supplier: Med Vet International DNX12
Dermabond Advanced is intended for topical application only to hold closed easily approximated skin edges of wounds from surgical incisions, including punctures from minimally invasive surgery, and simple, thoroughly cleansed, trauma-induced lacerations. Dermabond Advanced Adhesive is a wound closure device that adds strength when used in conjunction with deep dermal stitches after strength, provides a protective barrier and demonstrated in vitro inhibits bacteria. 12 per box.When used in addition to sutures, Dermabond Advanced Adhesive was shown ex vivo to add 75% more strength to wound closure than sutures alone. Dermabond Advanced Adhesive provides a microbial barrier with 99% protection in vitro for at least 72 hours against organisms commonly responsible for surgical site infections. Dermabond Advanced Adhesive is supported by substantially more Randomized Controlled Trials (RCTs) than the top six competitors combined.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.