Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MP Biomedicals™ Skim Milk Powder
A nonfat dry milk powder is produced from fresh skim milk. Skim Milk Powder is a component used in formulating animal research diets.
$102.70 - $389.00
Specifications
Product Type | Animal Diet |
---|---|
For Use With (Application) | Formulating research animal diet |
Storage Requirements | Room Temperature |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MP290288705
|
MP Biomedicals™
0290288705 |
5 lb. |
Each for $102.70
|
|
|||||
MP290288725
|
MP Biomedicals™
0290288725 |
25 lb. |
Each for $389.00
|
|
|||||
Description
- Skim milk powder
- Used in formulating research animal diets
- It is heated to achieve denaturation of the whey proteins
- Proteins, enzymes & peptides
Specifications
Animal Diet | |
Formulating research animal diet | |
Room Temperature |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title