Learn More
Pendotech Pendotech 1/8" Single Use Temperature Sensor
Supplier: Pendotech TEMPSN012
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Single use temperature sensor, non-sterile, 1/8" inlet/outlet, polysulfone