Learn More
Pendotech Pendotech 1/2 Inch Hose Barb Single Use Temperature Sensor
Supplier: Pendotech TEMPSN050
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Single Use Temperature Sensor with 1/2 inch hosebarb and polysulfone body