Learn More
Chemglass Life Sciences Manometer, Bubbler, Single Reservoir, Airfree, Schlenk
Supplier: Chemglass Life Sciences AF045302
Manometer, Bubbler, Single Reservoir, Airfree, Schlenk. Similar to the AF-0453-01 but with one reservoir. Overall dimensions are 50mm W x 1200mm H. PLEASE NOTE: NOT SUPPLIED WITH MERCURY.