Learn More
Pickering Laboratories Inc Simulated Sea Water for Permeation Testing, Not Stabilized, each (950 mL/bottle)
Supplier: Pickering Laboratories Inc 17000807
This solution was formulated according to Test Operations Procedure 08-2-501A that describes permeation testing of materials with chemical agents or simulants (Swatch Testing)