Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ORS Nasco Simple Green™ Automotive Cleaner/Degreaser
Safer alternative to toxic cleaners, solvents and bleaches
Supplier: ORS Nasco 67613012
This item is not returnable.
View return policy
Description
- Can be used on any washable, indoor, outdoor and automotive surface
- Cuts through brake dust, road tar, bug juice, and oil
- Non-abrasive
- Nontoxic, biodegradable formula
- Pump spray, 24 oz. (710mL)
- Clear
Specifications
Clear | |
Pump spray bottle | |
24 oz. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title