Learn More
HANNA INSTRUMENTS INC Silicone Oil (15 mL)
Supplier: HANNA INSTRUMENTS INC HI9870358
HI98703-58 silicone oil is for use with Hanna's line of turbidity meters. This easy to use dropper bottle ensures the glass cuvette used for turbidimetric measurements remains free from imperfections and scratches, helping to provide accurate readings every time.Convenient dropper bottleMarked with lot number and expiration dateLight tight bottle
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.