Learn More
Crescent Chemical Co Inc SILICONE SOLUTION SERVA FOR
Supplier: Crescent Chemical Co Inc SE35130.02
Silicone solution SERVA for siliconizing glass and metal in isopropanol Suitable for siliconizing UV quartz cuvettes. Size - 1L Storage Conditions - +15 °C TO +30 °C Catalog Number - 35130.02