Learn More
Crescent Chemical Co Inc SILICONE DC 550 FLUID; 115 CST
Supplier: Crescent Chemical Co Inc SE35145.01
Silicone DC 550 fluid; 115 cst pract. (Polyphenylmethyldimethylsiloxane)In terms of SI-units: 1 cst = 10-6m2s-1(25 % Methyl-75 % phenylsilicone). Excellent thermostability; lubricant; for instrument sterilization.Density (25 °C) 1.06 - 1.07 Size - 500G Storage Conditio