Learn More
Electron Microscopy Sciences Silicone Carbide Powder, 35 µm particles 1 LB
Supplier:  Electron Microscopy Sciences 5026035
Encompass_Preferred
Dark gray 98% SiC. Hardness, Moh's scale 9.0; Knoop Scale 2500. Crystallography: hexagonal–alpha and beta phase.
Size: 35 µm
Catalog No. 50-997-179