Learn More
Crescent Chemical Co Inc SILICONE ANTI-FOAM EMULSION
Supplier: Crescent Chemical Co Inc SE35119.02
Silicone anti-foam emulsion, 30 % Dow Corning 30 % polydimethylsiloxane in water; contains traces of emulsifiers derived from plant. Size - 500ML Storage Conditions - +15 C TO +30 C Catalog Number - 35119.02