Learn More
MilliporeSigma™ Supelco™ Silica PLOT Capillary GC Column
Porous-layer open-tubular (PLOT) columns are a powerful tool used in gas chromatography for analysis of fixed gases, light hydrocarbons and volatile solvents. They are mostly preferred over traditional columns.
Supplier: MilliporeSigma™ Supelco™ 28801U
Description
Application: This column can be used for the separation of C1-C4 hydrocarbons (even in the presence of moisture), fluorocarbons, sulfur gases, and carbon dioxide. USP Code: None. Phase:. Silica. Temp. Limits:. -80 C to 225 C (isothermal or programmed)Specifications
None | |
0.32 mm | |
Gas Chromatography (GC) | |
Fused Silica |
Capillary GC | |
-80°C to +225°C (isothermal or programmed) | |
30 m | |
Silica |